siRNA Details
| Gene Symbol: Trim80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033775.3 | NP_001028947.1 | 8 | 1867 | ||
| GenBank mRNA | AK133415.1 | - | 8 | 1868 | ||
| BC147608.1 | - | 8 | 1885 | 9056598 | ||
| BC157912.1 | - | 8 | 1885 | 9088129 | ||

Trim80
Genomic Map