siRNA Details
| Gene Symbol: Mfsd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033488.2 | NP_001028660.2 | 14 | 1884 | ||
| GenBank mRNA | AK133061.1 | - | 11 | 2786 | ||
| AK156138.1 | - | 15 | 2037 | |||
| AK157892.1 | - | 12 | 2567 | |||
| AK162620.1 | - | 12 | 1629 | |||
| AK171660.1 | - | 9 | 2248 | |||
| AK172423.1 | - | 14 | 1885 | |||

Mfsd2b
Genomic Map