siRNA Details
| Gene Symbol: Gm5447 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077314.1 | - | 1 | 1633 | ||
| BC147407.1 | - | 1 | 1590 | 9056483 | ||
| BC147408.1 | - | 1 | 1593 | 9056476 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5447| Gene Symbol: Gm5447 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077314.1 | - | 1 | 1633 | ||
| BC147407.1 | - | 1 | 1590 | 9056483 | ||
| BC147408.1 | - | 1 | 1593 | 9056476 | ||