siRNA Details
| Gene Symbol: Plcxd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134480.1 | NP_001127952.1 | 4 | 5019 | ||
| GenBank mRNA | AK086534.1 | - | 1 | 1280 | ||
| BC049364.1 | - | 1 | 2577 | |||
| BC064457.1 | - | 1 | 978 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plcxd2| Gene Symbol: Plcxd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134480.1 | NP_001127952.1 | 4 | 5019 | ||
| GenBank mRNA | AK086534.1 | - | 1 | 1280 | ||
| BC049364.1 | - | 1 | 2577 | |||
| BC064457.1 | - | 1 | 978 | |||