siRNA Details
Gene Symbol: Plcxd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134480.1 | NP_001127952.1 | 4 | 6654 | ||
GenBank mRNA | BC049364.1 | - | 1 | 4212 | ||
BC064457.1 | - | 1 | 2613 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Plcxd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134480.1 | NP_001127952.1 | 4 | 6654 | ||
GenBank mRNA | BC049364.1 | - | 1 | 4212 | ||
BC064457.1 | - | 1 | 2613 |