siRNA Details
| Gene Symbol: Spink11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048217.3 | NP_001041682.2 | 5 | 441 | ||
| GenBank mRNA | AK136741.1 | - | 5 | 439 | ||
| DQ437331.1 | - | 5 | 441 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spink11| Gene Symbol: Spink11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048217.3 | NP_001041682.2 | 5 | 441 | ||
| GenBank mRNA | AK136741.1 | - | 5 | 439 | ||
| DQ437331.1 | - | 5 | 441 | |||