siRNA Details
| Gene Symbol: Khdc1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033904.1 | NP_001029076.1 | 1 | 60 | ||
| GenBank mRNA | BC099381.1 | - | 1 | 60 | 30937206 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Khdc1c| Gene Symbol: Khdc1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033904.1 | NP_001029076.1 | 1 | 60 | ||
| GenBank mRNA | BC099381.1 | - | 1 | 60 | 30937206 | |