siRNA Details
Gene Symbol: Khdc1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033904.1 | NP_001029076.1 | 1 | 325 | ||
GenBank mRNA | BC099381.1 | - | 1 | 325 | 30937206 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Khdc1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033904.1 | NP_001029076.1 | 1 | 325 | ||
GenBank mRNA | BC099381.1 | - | 1 | 325 | 30937206 |