siRNA Details
| Gene Symbol: I830077J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033780.3 | NP_001028952.1 | 6 | 907 | ||
| GenBank mRNA | AK152510.1 | - | 6 | 910 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

I830077J02Rik| Gene Symbol: I830077J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033780.3 | NP_001028952.1 | 6 | 907 | ||
| GenBank mRNA | AK152510.1 | - | 6 | 910 | ||