siRNA Details
| Gene Symbol: Minos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163006.2 | NP_001156478.1 | 4 | 1495 | ||
| GenBank mRNA | AK030343.1 | - | 4 | 1448 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Minos1| Gene Symbol: Minos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163006.2 | NP_001156478.1 | 4 | 1495 | ||
| GenBank mRNA | AK030343.1 | - | 4 | 1448 | ||