siRNA Details
| Gene Symbol: Gm13154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014397.4 | NP_001014419.2 | 6 | 2527 | ||
| GenBank mRNA | BC085112.1 | - | 6 | 2513 | 30731985 | |
| Gene Symbol: Gm13152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC066875.1 | - | 2592 | 30111586 | ||

Gm13154
Genomic Map