siRNA Details
Gene Symbol: Gm13154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014397.4 | NP_001014419.2 | 6 | 602 | ||
GenBank mRNA | BC085112.1 | - | 6 | 584 | 30731985 | |
CF896923.1 | - | 4 | 478 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm13154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014397.4 | NP_001014419.2 | 6 | 602 | ||
GenBank mRNA | BC085112.1 | - | 6 | 584 | 30731985 | |
CF896923.1 | - | 4 | 478 |