siRNA Details
| Gene Symbol: Rnf207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033489.2 | NP_001028661.2 | 1 | 157 | ||
| GenBank mRNA | AK042379.1 | - | 1 | 140 | ||
| AK052224.1 | - | 1 | 158 | |||
| BC022631.1 | - | 1 | 151 | |||
| BC023223.1 | - | 1 | 13 | |||
| BC033405.1 | - | 1 | 506 | |||
| BC141256.1 | - | 1 | 41 | 9055868 | ||
| BC141257.1 | - | 1 | 41 | 9055869 | ||

Rnf207
Genomic Map