siRNA Details
| Gene Symbol: Nom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033457.2 | NP_001028629.2 | 1 | 857 | ||
| GenBank mRNA | AK148406.1 | - | 1 | 861 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Nom1
Nom1| Gene Symbol: Nom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033457.2 | NP_001028629.2 | 1 | 857 | ||
| GenBank mRNA | AK148406.1 | - | 1 | 861 | ||
 Back To Top
 Back To Top