siRNA Details
| Gene Symbol: Nom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033457.2 | NP_001028629.2 | 4 | 1476 | ||
| GenBank mRNA | AK148406.1 | - | 4 | 1480 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nom1| Gene Symbol: Nom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033457.2 | NP_001028629.2 | 4 | 1476 | ||
| GenBank mRNA | AK148406.1 | - | 4 | 1480 | ||