siRNA Details
Gene Symbol: Mn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081235.1 | NP_001074704.1 | 2 | 4757 | ||
GenBank mRNA | AK142207.1 | - | 3869 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081235.1 | NP_001074704.1 | 2 | 4757 | ||
GenBank mRNA | AK142207.1 | - | 3869 |