siRNA Details
| Gene Symbol: Hpcal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017357.1 | NP_059053.1 | 2 | 218 | ||
| XM_006238829.2 | XP_006238891.1 | 3 | 513 | |||
| GenBank mRNA | D13125.1 | - | 2 | 218 | ||
| D14819.1 | - | 646 | ||||
| FQ212482.1 | - | 3 | 346 | |||

Hpcal4
Genomic Map