siRNA Details
| Gene Symbol: Gm5591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013810.2 | NP_001013832.1 | 3 | 3028 | ||
| GenBank mRNA | AK029937.1 | - | 3 | 3029 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5591| Gene Symbol: Gm5591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013810.2 | NP_001013832.1 | 3 | 3028 | ||
| GenBank mRNA | AK029937.1 | - | 3 | 3029 | ||