siRNA Details
| Gene Symbol: Trim34b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243916.1 | NP_001230845.1 | 5, 6 | 944 | ||
| GenBank mRNA | BC145515.1 | - | 5, 6 | 944 | ||
| Gene Symbol: Trim34a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF220140.1 | - | 1103 | |||
| AK089187.1 | - | 1218 | ||||
| AK171840.1 | - | 6, 7 | 1163 | |||
| AK172135.1 | - | 6, 7 | 1163 | |||

Trim34b
Genomic Map