siRNA Details
Gene Symbol: Sult2a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184980.1 | NP_001171909.1 | 2, 3 | 431 | ||
GenBank mRNA | AI315523.1 | - | 2, 3 | 431 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sult2a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184980.1 | NP_001171909.1 | 2, 3 | 431 | ||
GenBank mRNA | AI315523.1 | - | 2, 3 | 431 |