siRNA Details
| Gene Symbol: Ccdc36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135198.1 | NP_001128670.1 | 7 | 1053 | ||
| GenBank mRNA | BC058624.1 | - | 7 | 1058 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc36| Gene Symbol: Ccdc36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135198.1 | NP_001128670.1 | 7 | 1053 | ||
| GenBank mRNA | BC058624.1 | - | 7 | 1058 | ||