siRNA Details
| Gene Symbol: Smim9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033786.2 | NP_001028958.1 | 5 | 649 | ||
| GenBank mRNA | AK132828.1 | - | 5 | 653 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smim9| Gene Symbol: Smim9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033786.2 | NP_001028958.1 | 5 | 649 | ||
| GenBank mRNA | AK132828.1 | - | 5 | 653 | ||