siRNA Details
| Gene Symbol: Smim9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033786.2 | NP_001028958.1 | 3 | 336 | ||
| GenBank mRNA | AK132828.1 | - | 3 | 340 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smim9| Gene Symbol: Smim9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033786.2 | NP_001028958.1 | 3 | 336 | ||
| GenBank mRNA | AK132828.1 | - | 3 | 340 | ||