siRNA Details
Gene Symbol: Shisa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207021.1 | NP_001193950.1 | 4 | 1370 | ||
GenBank mRNA | BG919271.1 | - | 453 | |||
BU558590.1 | - | 4 | 577 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Shisa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207021.1 | NP_001193950.1 | 4 | 1370 | ||
GenBank mRNA | BG919271.1 | - | 453 | |||
BU558590.1 | - | 4 | 577 |