siRNA Details
| Gene Symbol: Serpinb6e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045535.2 | NP_001039000.2 | 7 | 879 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpinb6e| Gene Symbol: Serpinb6e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045535.2 | NP_001039000.2 | 7 | 879 | ||