siRNA Details
| Gene Symbol: Rabep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019124.1 | NP_061997.1 | 2, 3 | 219 | ||
| XM_006246795.2 | XP_006246857.1 | 2, 3 | 382 | |||
| XM_006246796.2 | XP_006246858.1 | 2, 3 | 382 | |||
| XM_008767861.1 | XP_008766083.1 | 2, 3 | 1013 | |||
| GenBank mRNA | D85844.1 | - | 2, 3 | 219 | ||
| U70777.1 | - | 2, 3 | 366 | |||

Rabep1
Genomic Map