siRNA Details
Gene Symbol: Ldlrad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033979.1 | NP_001029151.1 | 4 | 1779 | ||
GenBank mRNA | AK132881.1 | - | 4 | 1779 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ldlrad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033979.1 | NP_001029151.1 | 4 | 1779 | ||
GenBank mRNA | AK132881.1 | - | 4 | 1779 |