siRNA Details
| Gene Symbol: Slc2a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085529.1 | NP_001078998.1 | 12 | 1425 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc2a7| Gene Symbol: Slc2a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085529.1 | NP_001078998.1 | 12 | 1425 | ||