siRNA Details
| Gene Symbol: Fam92b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033980.2 | NP_001029152.2 | 9 | 1175 | ||
| GenBank mRNA | AK131790.1 | - | 9 | 1175 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam92b| Gene Symbol: Fam92b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033980.2 | NP_001029152.2 | 9 | 1175 | ||
| GenBank mRNA | AK131790.1 | - | 9 | 1175 | ||