siRNA Details
| Gene Symbol: Gm5766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030194.1 | - | 1 | 987 | ||
| BC147642.1 | - | 1 | 346 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5766| Gene Symbol: Gm5766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030194.1 | - | 1 | 987 | ||
| BC147642.1 | - | 1 | 346 | |||