siRNA Details
| Gene Symbol: Tmem141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040130.3 | NP_001035219.2 | 5 | 748 | ||
| NM_001109993.1 | NP_001103463.1 | 4 | 1010 | |||
| GenBank mRNA | BY648636.1 | - | 1 | 387 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem141| Gene Symbol: Tmem141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040130.3 | NP_001035219.2 | 5 | 748 | ||
| NM_001109993.1 | NP_001103463.1 | 4 | 1010 | |||
| GenBank mRNA | BY648636.1 | - | 1 | 387 | ||