siRNA Details
| Gene Symbol: Atxn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080930.1 | NP_001074399.1 | 4 | 6045 | ||
| GenBank mRNA | AK082766.1 | - | 1 | 342 | ||
| BC038886.1 | - | 1 | 1622 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atxn1l| Gene Symbol: Atxn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080930.1 | NP_001074399.1 | 4 | 6045 | ||
| GenBank mRNA | AK082766.1 | - | 1 | 342 | ||
| BC038886.1 | - | 1 | 1622 | |||