siRNA Details
| Gene Symbol: Sco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040026.1 | NP_001035115.1 | 6 | 3700 | ||
| GenBank mRNA | AK018523.1 | - | 1 | 1907 | ||
| AK083729.1 | - | 2122 | ||||
| BC038283.1 | - | 4 | 2461 | |||
| BC139008.1 | - | 6 | 3697 | 8862030 | ||
| BC139009.1 | - | 6 | 3698 | 8862031 | ||

Sco1
Genomic Map