siRNA Details
Gene Symbol: BB287469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177573.1 | NP_001171044.1 | 2 | 1314 | ||
GenBank mRNA | AK139752.1 | - | 2 | 1314 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BB287469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177573.1 | NP_001171044.1 | 2 | 1314 | ||
GenBank mRNA | AK139752.1 | - | 2 | 1314 |