siRNA Details
| Gene Symbol: BB287469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177573.1 | NP_001171044.1 | 2 | 1679 | ||
| GenBank mRNA | AK139752.1 | - | 2 | 1679 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BB287469| Gene Symbol: BB287469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177573.1 | NP_001171044.1 | 2 | 1679 | ||
| GenBank mRNA | AK139752.1 | - | 2 | 1679 | ||