siRNA Details
| Gene Symbol: Gm5797 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025085.2 | NP_001020256.1 | 1 | 37 | ||
| GenBank mRNA | BC090991.1 | - | 1 | 37 | 6742820 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5797| Gene Symbol: Gm5797 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025085.2 | NP_001020256.1 | 1 | 37 | ||
| GenBank mRNA | BC090991.1 | - | 1 | 37 | 6742820 | |