siRNA Details
| Gene Symbol: Wdfy4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146022.2 | NP_001139494.1 | 56, 57 | 9024 | ||
| GenBank mRNA | AK155186.1 | - | 17, 18 | 2164 | ||
| AK173214.1 | - | 5, 6 | 802 | |||
| DT911337.1 | - | 13, 14 | 1733 | |||

Wdfy4
Genomic Map