siRNA Details
Gene Symbol: Gm5803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165971.1 | NP_001159443.1 | 1 | 33 | ||
GenBank mRNA | BC157911.1 | - | 1 | 33 | ||
BC172092.1 | - | 1 | 33 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165971.1 | NP_001159443.1 | 1 | 33 | ||
GenBank mRNA | BC157911.1 | - | 1 | 33 | ||
BC172092.1 | - | 1 | 33 |