siRNA Details
| Gene Symbol: Matr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019149.3 | NP_062022.2 | 2 | 379 | ||
| XM_006254534.2 | XP_006254596.1 | 2 | 401 | |||
| XM_006254535.2 | XP_006254597.1 | 2 | 352 | |||
| GenBank mRNA | AB205483.1 | - | 1 | 1149 | ||
| BC062231.1 | - | 2 | 402 | |||
| BC168723.1 | - | 2 | 351 | |||
| M63485.1 | - | 2 | 364 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010771, BC029070, BC090833, CT010322, AK087939, and others.

Matr3
Genomic Map