siRNA Details
| Gene Symbol: Gm5803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165971.1 | NP_001159443.1 | 1 | 399 | ||
| GenBank mRNA | BC157911.1 | - | 1 | 399 | ||
| BC172092.1 | - | 1 | 399 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5803| Gene Symbol: Gm5803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165971.1 | NP_001159443.1 | 1 | 399 | ||
| GenBank mRNA | BC157911.1 | - | 1 | 399 | ||
| BC172092.1 | - | 1 | 399 | |||