siRNA Details
| Gene Symbol: Fam186b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081254.1 | NP_001074723.1 | 8 | 2919 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV261348.1 | - | 387 | Chr. - - [-] on Build | ||

Fam186b
Genomic Map