siRNA Details
| Gene Symbol: Gm2889 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK145513.1 | - | 2311 | |||
| AK166853.1 | - | 2311 | ||||
| Gene Symbol: G430049J08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144159.1 | - | 1 | 2335 | ||
| AK166742.1 | - | 1 | 2331 | |||

Gm2889
Genomic Map