siRNA Details
| Gene Symbol: G430049J08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144159.1 | - | 1 | 1768 | ||
| AK166742.1 | - | 1 | 1764 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

G430049J08Rik| Gene Symbol: G430049J08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144159.1 | - | 1 | 1768 | ||
| AK166742.1 | - | 1 | 1764 | |||