siRNA Details
| Gene Symbol: Gm16432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034899.3 | NP_001030071.2 | 8 | 778 | ||
| GenBank mRNA | BC147679.1 | - | 8 | 778 | 9056652 | |
| BC147686.1 | - | 8 | 778 | 9056659 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16432| Gene Symbol: Gm16432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034899.3 | NP_001030071.2 | 8 | 778 | ||
| GenBank mRNA | BC147679.1 | - | 8 | 778 | 9056652 | |
| BC147686.1 | - | 8 | 778 | 9056659 | ||