siRNA Details
| Gene Symbol: Scrt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160410.1 | NP_001153882.1 | 2 | 1844 | ||
| GenBank mRNA | AK090280.1 | - | 1 | 894 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scrt2| Gene Symbol: Scrt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160410.1 | NP_001153882.1 | 2 | 1844 | ||
| GenBank mRNA | AK090280.1 | - | 1 | 894 | ||