siRNA Details
| Gene Symbol: Aqp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019157.2 | NP_062030.2 | 5 | 648 | ||
| GenBank mRNA | AB000507.2 | - | 5 | 648 | ||
| AY120935.1 | - | 4 | 387 | |||
| AY157737.1 | - | 4 | 387 | |||
| BC072695.1 | - | 5 | 608 | 7104684 | ||

Aqp7
Genomic Map