siRNA Details
| Gene Symbol: I0C0044D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK145435.1 | - | 3 | 1948 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

I0C0044D17Rik| Gene Symbol: I0C0044D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK145435.1 | - | 3 | 1948 | ||