siRNA Details
| Gene Symbol: Smok3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168602.1 | NP_001162073.1 | 2 | 2007 | ||
| GenBank mRNA | AJ245454.1 | - | 2 | 2007 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smok3a| Gene Symbol: Smok3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168602.1 | NP_001162073.1 | 2 | 2007 | ||
| GenBank mRNA | AJ245454.1 | - | 2 | 2007 | ||