siRNA Details
| Gene Symbol: Gm5878 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034902.2 | NP_001030074.2 | 3 | 1263 | ||
| GenBank mRNA | AK132708.1 | - | 3 | 1263 | ||
| BC150951.1 | - | 3 | 1190 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5878| Gene Symbol: Gm5878 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034902.2 | NP_001030074.2 | 3 | 1263 | ||
| GenBank mRNA | AK132708.1 | - | 3 | 1263 | ||
| BC150951.1 | - | 3 | 1190 | |||