siRNA Details
| Gene Symbol: Scgb1b20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270543.1 | NP_001257472.1 | 3 | 313 | ||
| GenBank mRNA | CK616096.1 | - | 3 | 314 | ||
| CK616412.1 | - | 3 | 309 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scgb1b20| Gene Symbol: Scgb1b20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270543.1 | NP_001257472.1 | 3 | 313 | ||
| GenBank mRNA | CK616096.1 | - | 3 | 314 | ||
| CK616412.1 | - | 3 | 309 | |||